Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036110.1 | NP_001031187.2 | 10 - 11 | 1082 | 108 | |
NM_001198292.1 | NP_001185221.1 | 10 - 11 | 1017 | 108 | ||
NM_001333642.1 | NP_001319224.1 | 9 - 10 | 1169 | 108 | ||
NM_001333643.1 | NP_001322136.1 | 9 - 10 | 1213 | 108 | ||
NM_104314.6 | NP_175838.4 | 10 - 11 | 1145 | 108 | ||
NM_202295.2 | NP_974024.2 | 9 - 10 | 1198 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |