Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333652.1 | NP_001321327.1 | 13 - 14 | 1871 | 81 | |
NM_104327.4 | NP_175851.1 | 13 - 14 | 1666 | 81 | ||
GenBank mRNA | AF286718.1 | - | 13 - 14 | 1585 | 81 | |
AY064012.1 | - | 13 - 14 | 1582 | 81 | ||
AY091411.1 | - | 13 - 14 | 1475 | 81 | ||
BT026022.1 | - | 13 - 14 | 1561 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |