Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333652.1 | NP_001321327.1 | 16 - 17 | 2120 | 123 | |
NM_104327.4 | NP_175851.1 | 16 - 17 | 1915 | 123 | ||
GenBank mRNA | AF286718.1 | - | 16 - 17 | 1834 | 123 | |
AY064012.1 | - | 16 - 17 | 1831 | 123 | ||
AY091411.1 | - | 16 - 17 | 1724 | 123 | ||
BT026022.1 | - | 16 - 17 | 1810 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |