Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG18H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104346.3 | NP_564664.1 | 1 - 2 | 623 | 125 | |
GenBank mRNA | AY142626.1 | - | 1 - 2 | 200 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |