Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104376.3 | NP_175900.1 | 4 - 5 | 1291 | 85 | |
GenBank mRNA | AY093104.1 | - | 4 - 5 | 1235 | 85 | |
BT010358.1 | - | 4 - 5 | 1147 | 85 | ||
L04637.1 | - | 4 - 5 | 1227 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |