Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104376.3 | NP_175900.1 | 5 - 6 | 1403 | 121 | |
GenBank mRNA | AY093104.1 | - | 5 - 6 | 1347 | 121 | |
BT010358.1 | - | 5 - 6 | 1259 | 121 | ||
L04637.1 | - | 5 - 6 | 1339 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |