Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCL33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036118.4 | NP_001031195.4 | 3 - 4 | 315 | 71 | |
NM_001333699.1 | NP_001319238.1 | 3 - 4 | 548 | 71 | ||
NM_104406.6 | NP_564685.4 | 3 - 4 | 682 | 71 | ||
GenBank mRNA | AF099940.2 | - | 3 - 4 | 285 | 71 | |
AJ293799.1 | - | 3 - 4 | 283 | 71 | ||
BT020420.1 | - | 2 - 3 | 224 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Arabidopsis thaliana Probe spans exons |