Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084254.1 | NP_001077723.1 | 13 - 14 | 2078 | 104 | |
NM_104408.5 | NP_175929.3 | 13 - 14 | 2341 | 104 | ||
GenBank mRNA | AY250845.1 | - | 13 - 14 | 2066 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |