Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084254.1 | NP_001077723.1 | 1 - 2 | 275 | 64 | |
NM_104408.5 | NP_175929.3 | 1 - 2 | 542 | 64 | ||
GenBank mRNA | AY250845.1 | - | 1 - 2 | 267 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |