Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198311.2 | NP_001185240.1 | 3 - 4 | 523 | 100 | |
NM_001198312.2 | NP_001185241.1 | 3 - 4 | 523 | 100 | ||
NM_001333713.1 | NP_001321787.1 | 3 - 4 | 523 | 100 | ||
NM_001333714.1 | NP_001321789.1 | 3 - 4 | 523 | 100 | ||
NM_001333715.1 | NP_001321788.1 | 2 - 3 | 698 | 100 | ||
NM_104425.1 | NP_564692.1 | 2 - 3 | 671 | 100 | ||
GenBank mRNA | AY050425.1 | - | 2 - 3 | 671 | 100 | |
BT002240.1 | - | 1 - 2 | 120 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |