Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198312.2 | NP_001185241.1 | 8 - 9 | 1997 | 73 | |
NM_001333714.1 | NP_001321789.1 | 8 - 9 | 2036 | 73 | ||
NM_001333715.1 | NP_001321788.1 | 7 - 8 | 2211 | 73 | ||
NM_104425.1 | NP_564692.1 | 7 - 8 | 2145 | 73 | ||
GenBank mRNA | AY050425.1 | - | 7 - 8 | 2145 | 73 | |
BT002240.1 | - | 6 - 7 | 1594 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |