Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104436.2 | NP_175956.1 | 4 - 5 | 983 | 71 | |
GenBank mRNA | AY071851.1 | - | 4 - 5 | 982 | 71 | |
DQ446362.1 | - | 4 - 5 | 982 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |