Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036119.2 | NP_001031196.1 | 5 - 6 | 758 | 68 | |
NM_104449.3 | NP_175969.2 | 5 - 6 | 665 | 68 | ||
GenBank mRNA | AK317482.1 | - | 5 - 6 | 640 | 68 | |
AY136416.1 | - | 5 - 6 | 647 | 68 | ||
BT006591.1 | - | 4 - 5 | 393 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |