Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104450.4 | NP_175970.1 | 3 - 4 | 598 | 88 | |
GenBank mRNA | AK227977.1 | - | 3 - 4 | 620 | 88 | |
BT004640.1 | - | 3 - 4 | 527 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |