Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104479.2 | NP_175997.1 | 2 - 3 | 617 | 126 | |
NM_179486.2 | NP_849817.1 | 1 - 2 | 1521 | 126 | ||
GenBank mRNA | AF198054.1 | - | 2 - 3 | 568 | 126 | |
AK228347.1 | - | 1 - 2 | 592 | 126 | ||
AY085996.1 | - | 2 - 3 | 577 | 126 | ||
BT005873.1 | - | 1 - 2 | 279 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |