Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DI19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104507.4 | NP_564715.4 | 1 - 2 | 197 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |