Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DI19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333775.1 | NP_001319253.1 | 5 - 6 | 1063 | 130 | |
NM_104507.4 | NP_564715.4 | 5 - 6 | 817 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |