Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104523.4 | NP_176040.1 | 3 - 4 | 493 | 151 | |
GenBank mRNA | AF378880.1 | - | 3 - 4 | 232 | 151 | |
AK221762.1 | - | 3 - 4 | 243 | 151 | ||
AY045596.1 | - | 3 - 4 | 209 | 151 | ||
AY052739.1 | - | 2 - 3 | 156 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |