Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198327.1 | NP_001185256.1 | 13 - 14 | 1623 | 75 | |
NM_001333808.1 | NP_001319262.1 | 12 - 13 | 1510 | 75 | ||
NM_001333809.1 | NP_001320412.1 | 13 - 14 | 1916 | 75 | ||
NM_104580.5 | NP_176096.1 | 13 - 14 | 1853 | 75 | ||
GenBank mRNA | BT002018.1 | - | 13 - 14 | 1626 | 75 | |
BT008861.1 | - | 11 - 12 | 1175 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |