Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036129.3 | NP_001031206.1 | 6 - 7 | 790 | 96 | |
NM_001198330.1 | NP_001185259.1 | 7 - 8 | 696 | 96 | ||
NM_001333816.1 | NP_001320703.1 | 7 - 8 | 820 | 96 | ||
NM_001333817.1 | NP_001320705.1 | 6 - 7 | 776 | 96 | ||
NM_001333818.1 | NP_001320704.1 | 5 - 6 | 486 | 96 | ||
NM_104599.5 | NP_176114.2 | 7 - 8 | 820 | 96 | ||
NM_179492.4 | NP_849823.1 | 7 - 8 | 758 | 96 | ||
GenBank mRNA | AK316811.1 | - | 6 - 7 | 672 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |