Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CW7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333852.1 | NP_001319271.1 | 4 - 5 | 710 | 74 | |
NM_001333853.1 | NP_001321061.1 | 4 - 5 | 710 | 74 | ||
NM_001333854.1 | NP_001321060.1 | 4 - 5 | 710 | 74 | ||
NM_104645.4 | NP_564743.1 | 4 - 5 | 710 | 74 | ||
NM_179496.2 | NP_849827.1 | 4 - 5 | 695 | 74 | ||
NM_179497.1 | NP_849828.1 | 4 - 5 | 608 | 74 | ||
GenBank mRNA | AB028226.1 | - | 4 - 5 | 515 | 74 | |
AK227804.1 | - | 4 - 5 | 592 | 74 | ||
BT004825.1 | - | 4 - 5 | 403 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |