Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTU16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104662.4 | NP_176178.1 | 1 - 2 | 510 | 61 | |
GenBank mRNA | BT014880.1 | - | 1 - 2 | 329 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |