Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036131.3 | NP_001031208.1 | 2 - 3 | 152 | 68 | |
NM_001198337.1 | NP_001185266.1 | 2 - 3 | 398 | 68 | ||
NM_104668.4 | NP_176184.1 | 2 - 3 | 348 | 68 | ||
NM_179499.2 | NP_849830.1 | 2 - 3 | 303 | 68 | ||
GenBank mRNA | AK317214.1 | - | 2 - 3 | 108 | 68 | |
BT002748.1 | - | 1 - 2 | 37 | 68 | ||
U83245.1 | - | 2 - 3 | 151 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |