Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104675.3 | NP_176191.1 | 14 - 15 | 1658 | 96 | |
GenBank mRNA | AY091777.1 | - | 14 - 15 | 1635 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |