Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104675.3 | NP_176191.1 | 20 - 21 | 2459 | 106 | |
GenBank mRNA | AY091777.1 | - | 20 - 21 | 2436 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |