Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333876.1 | NP_001322340.1 | 2 - 3 | 220 | 97 | |
NM_104701.3 | NP_176217.2 | 2 - 3 | 197 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |