Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104731.5 | NP_176247.2 | 3 - 4 | 445 | 133 | |
GenBank mRNA | AK229327.1 | - | 3 - 4 | 428 | 133 | |
BT011005.1 | - | 3 - 4 | 295 | 133 | ||
BT015384.1 | - | 3 - 4 | 257 | 133 |
Gene Symbol: FLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK230362.1 | - | 890 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |