Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRPAC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333919.1 | NP_001320360.1 | 1 - 2 | 394 | 99 | |
NM_104766.5 | NP_176282.1 | 2 - 3 | 288 | 99 | ||
NM_179502.4 | NP_849833.1 | 2 - 3 | 277 | 99 | ||
NM_202328.3 | NP_974057.1 | 1 - 2 | 293 | 99 | ||
GenBank mRNA | AK226775.1 | - | 2 - 3 | 242 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |