Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RECQ4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104773.7 | NP_176289.7 | 4 - 5 | 486 | 110 | |
GenBank mRNA | AJ404474.1 | - | 4 - 5 | 486 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |