Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF73-I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036138.2 | NP_001031215.1 | 1 - 2 | 249 | 61 | |
NM_001333924.1 | NP_001319278.1 | 1 - 2 | 249 | 61 | ||
NM_001333925.1 | NP_001321253.1 | 1 - 2 | 243 | 61 | ||
NM_104782.4 | NP_176297.1 | 1 - 2 | 243 | 61 | ||
NM_179504.2 | NP_849835.1 | 1 - 2 | 249 | 61 | ||
GenBank mRNA | AK316692.1 | - | 1 - 2 | 186 | 61 | |
AK316794.1 | - | 1 - 2 | 214 | 61 | ||
AY140900.1 | - | 1 - 2 | 215 | 61 | ||
AY150478.1 | - | 1 - 2 | 144 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |