Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104794.3 | NP_176308.2 | 4 - 5 | 589 | 90 | |
GenBank mRNA | DQ056504.1 | - | 4 - 5 | 546 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |