Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104875.3 | NP_176386.2 | 4 - 5 | 1079 | 104 | |
GenBank mRNA | DQ446385.1 | - | 4 - 5 | 1051 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |