Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104914.2 | NP_564793.2 | 4 - 5 | 871 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |