Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104916.4 | NP_176426.1 | 2 - 3 | 1021 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Arabidopsis thaliana Probe spans exons |