Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104923.4 | NP_176433.2 | 7 - 8 | 1143 | 73 | |
GenBank mRNA | X94306.1 | - | 1074 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |