Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BETA-VPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104948.4 | NP_176458.1 | 6 - 7 | 987 | 118 | |
GenBank mRNA | AF367254.1 | - | 6 - 7 | 891 | 118 | |
AY059156.1 | - | 6 - 7 | 861 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |