Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198369.1 | NP_001185298.1 | 15 - 16 | 3603 | 120 | |
NM_001334061.1 | NP_001320709.1 | 14 - 15 | 3793 | 120 | ||
NM_001334062.1 | NP_001320708.1 | 15 - 16 | 3603 | 120 | ||
NM_001334063.1 | NP_001320710.1 | 6 - 7 | 2690 | 120 | ||
NM_104980.4 | NP_176490.2 | 14 - 15 | 3768 | 120 | ||
GenBank mRNA | AY826515.1 | - | 13 - 14 | 3494 | 120 | |
DQ020657.1 | - | 14 - 15 | 3768 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |