Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125496.2 | NP_001118968.1 | 11 - 12 | 5091 | 123 | |
NM_001160750.1 | NP_001154222.1 | 11 - 12 | 5091 | 123 | ||
NM_001340760.1 | NP_001329658.1 | 11 - 12 | 5099 | 123 | ||
NM_001340762.1 | NP_001329652.1 | 11 - 12 | 5201 | 123 | ||
NM_001340763.1 | NP_001329653.1 | 10 - 11 | 5320 | 123 | ||
NM_001340764.1 | NP_001329654.1 | 11 - 12 | 5153 | 123 | ||
NM_001340765.1 | NP_001329655.1 | 11 - 12 | 5153 | 123 | ||
NM_001340766.1 | NP_001329656.1 | 11 - 12 | 5153 | 123 | ||
NM_001340767.1 | NP_001329657.1 | 11 - 12 | 5166 | 123 | ||
NM_117272.3 | NP_192939.2 | 11 - 12 | 5091 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |