Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340768.1 | NP_001319909.1 | 4 - 5 | 1076 | 73 | |
NM_001340769.1 | NP_001328579.1 | 3 - 4 | 1146 | 73 | ||
NM_001340770.1 | NP_001328580.1 | 4 - 5 | 1068 | 73 | ||
NM_117273.3 | NP_567385.1 | 1 - 2 | 658 | 73 | ||
NM_202809.3 | NP_974538.1 | 5 - 6 | 998 | 73 | ||
GenBank mRNA | AY050449.1 | - | 5 - 6 | 847 | 73 | |
AY097358.1 | - | 1 - 2 | 422 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |