Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129010.4 | NP_181004.2 | 1 - 2 | 295 | 70 | |
GenBank mRNA | AY102549.1 | - | 1 - 2 | 295 | 70 | |
AY954813.1 | - | 1 - 2 | 194 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |