Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129018.4 | NP_181011.2 | 5 - 6 | 1412 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |