Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202746.1 | NP_001189675.1 | 3 - 4 | 812 | 126 | |
NM_129020.3 | NP_181013.1 | 2 - 3 | 1077 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |