Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336523.1 | NP_001324697.1 | 15 - 16 | 2753 | 65 | |
NM_129022.8 | NP_181015.7 | 15 - 16 | 2564 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |