Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084535.1 | NP_001078004.1 | 3 - 4 | 361 | 84 | |
NM_129039.4 | NP_181032.1 | 3 - 4 | 478 | 84 | ||
GenBank mRNA | AY099664.1 | - | 3 - 4 | 353 | 84 | |
AY128851.1 | - | 3 - 4 | 341 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |