Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KUP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036407.2 | NP_001031484.1 | 6 - 7 | 1189 | 74 | |
NM_001336538.1 | NP_001324455.1 | 5 - 6 | 1861 | 74 | ||
NM_001336539.1 | NP_001324456.1 | 6 - 7 | 1199 | 74 | ||
NM_129058.5 | NP_181051.1 | 6 - 7 | 1314 | 74 | ||
GenBank mRNA | AK227188.1 | - | 6 - 7 | 1078 | 74 | |
AY099600.1 | - | 6 - 7 | 1074 | 74 | ||
BT002147.1 | - | 5 - 6 | 939 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |