Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336552.1 | NP_001323587.1 | 10 - 11 | 667 | 99 | |
NM_129086.4 | NP_181077.3 | 14 - 15 | 1227 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |