Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036145.3 | NP_001031222.1 | 1 - 2 | 383 | 96 | |
NM_001334075.1 | NP_001320678.1 | 1 - 2 | 360 | 96 | ||
NM_001334076.1 | NP_001320677.1 | 1 - 2 | 402 | 96 | ||
NM_001334077.1 | NP_001320676.1 | 2 - 3 | 301 | 96 | ||
NM_105005.5 | NP_176515.3 | 1 - 2 | 383 | 96 | ||
NM_202348.3 | NP_974077.1 | 1 - 2 | 383 | 96 | ||
GenBank mRNA | BT011569.1 | - | 1 - 2 | 244 | 96 | |
BT012253.1 | - | 1 - 2 | 58 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |