Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036145.3 | NP_001031222.1 | 4 - 5 | 601 | 78 | |
NM_001334075.1 | NP_001320678.1 | 4 - 5 | 578 | 78 | ||
NM_001334076.1 | NP_001320677.1 | 4 - 5 | 620 | 78 | ||
NM_001334077.1 | NP_001320676.1 | 5 - 6 | 519 | 78 | ||
NM_105005.5 | NP_176515.3 | 4 - 5 | 601 | 78 | ||
NM_202348.3 | NP_974077.1 | 4 - 5 | 601 | 78 | ||
GenBank mRNA | BT011569.1 | - | 4 - 5 | 462 | 78 | |
BT012253.1 | - | 4 - 5 | 276 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |