Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036145.3 | NP_001031222.1 | 7 - 8 | 783 | 91 | |
NM_001334075.1 | NP_001320678.1 | 7 - 8 | 760 | 91 | ||
NM_001334076.1 | NP_001320677.1 | 7 - 8 | 802 | 91 | ||
NM_001334077.1 | NP_001320676.1 | 8 - 9 | 701 | 91 | ||
NM_105005.5 | NP_176515.3 | 7 - 8 | 783 | 91 | ||
NM_202348.3 | NP_974077.1 | 7 - 8 | 783 | 91 | ||
GenBank mRNA | BT011569.1 | - | 7 - 8 | 644 | 91 | |
BT012253.1 | - | 7 - 8 | 458 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |