Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105046.3 | NP_564815.1 | 13 - 14 | 1980 | 105 | |
NM_202353.1 | NP_974082.1 | 13 - 14 | 1614 | 105 | ||
GenBank mRNA | AY062807.1 | - | 13 - 14 | 1567 | 105 | |
BT008895.1 | - | 13 - 14 | 1502 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |